missing translation for 'onlineSavingsMsg'
Learn More

Octamethylcyclotetrasilazane, 97%, Thermo Scientific Chemicals

Catalog Number 11339235
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1020-84-4
C8H28N4Si4
292.68
MFCD00046123
FIADVASZMLCQIF-UHFFFAOYSA-N
2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetrazatetrasilocane
C[Si]1(C)N[Si](C)(C)N[Si](C)(C)N[Si](C)(C)N1
This item is not returnable. View return policy
1020-84-4
C8H28N4Si4
292.68
MFCD00046123
FIADVASZMLCQIF-UHFFFAOYSA-N
2,2,4,4,6,6,8,8-octamethyl-1,3,5,7,2,4,6,8-tetrazatetrasilocane
C[Si]1(C)N[Si](C)(C)N[Si](C)(C)N[Si](C)(C)N1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 224°C to 225°C
Melting Point 94°C to 98°C
Quantity 5 g
UN Number UN2923
Beilstein 976248
Flash Point 66°C (150°F)
Formula Weight 292.68
Percent Purity 97%
Odor Ammonia-like
Sensitivity Moisture sensitive
Chemical Name or Material Octamethylcyclotetrasilazane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'