missing translation for 'onlineSavingsMsg'
Learn More

Octakis(trimethylsiloxy)silsesquioxane

Product Code. 11359748
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Unit Size:
100g
1g
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11359748 25 g 25g
11460690 1 g 1g
11369748 5 g 5g
11349748 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11359748 Supplier Thermo Scientific Alfa Aesar Supplier No. 044231.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 51777-38-9 | C24H72O10Si11 | 829.765 g/mol

Used as flavor and fragrance intermediates. It is an important raw material used intermediate in pharmaceuticals, organic synthesis, dye stuffs and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51777-38-9
Molecular Formula C24H72O10Si11
Molecular Weight (g/mol) 829.765
MDL Number MFCD01310212
InChI Key VLEKPQBXDHFSQO-UHFFFAOYSA-N
Synonym octakis trimethylsiloxy si-lsesquioxane, octakis trimethylsiloxy-t8-silsesquioxane
PubChem CID 71306897
IUPAC Name bis(trimethylsilyl) bis[tris(trimethylsilyloxy)silyl] silicate
SMILES C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C

Specifications

Color White
Quantity 25 g
Solubility Information Sparingly Soluble (0.61g/L) (25°C),Calc.
Formula Weight 1130.18
Physical Form Powder
Chemical Name or Material Octakis (trimethylsiloxy)silsesquioxane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.