missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octahydroindoline-2-carboxylic Acid Benzyl Ester (-)-Dibenzoyl-L-tartaric Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Octahydroindoline-2-carboxylic Acid Benzyl Ester (-)-Dibenzoyl-L-tartaric Acid |
| Analyte or Component Names | Octahydroindoline-2-carboxylic Acid Benzyl Ester (-)-Dibenzoyl-L-tartaric Acid |
| Shipping Condition | Room Temperature |
| Molecular Formula | C34H35NO10 |
| InChI Formula | InChI=1S/C18H14O8.C16H21NO2/c19-15(20)13(25-17(23)11-7-3-1-4-8-11)14(16(21)22)26-18(24)12-9-5-2-6-10-12;18-16(19-11-12-6-2-1-3-7-12)15-10-13-8-4-5-9-14(13)17-15/h1-10,13-14H,(H,19,20)(H,21,22);1-3,6-7,13-15,17H,4-5,8-11H2 |
| Solution Type | Neat |
| SMILES | O=C(C1CC2C(CCCC2)N1)OCC3=CC=CC=C3.O=C([C@H](OC(C4=CC=CC=C4)=O)[C@H](C(O)=O)OC(C5=CC=CC=C5)=O)O |
| IUPAC Name | benzyl octahydro-1H-indole-2-carboxylate (2R,3R)-2,3-bis(benzoyloxy)succinate |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 617.64 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?