missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Octagonal Shape PTFE Magnetic Stirring Bars

PTFE construction. Angular surface creates high turbulence even at low speeds.

Brand:  BRAND™ 137618

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10191631

  • £70.00 / Pack of 10
Estimated Shipment: 26-06-2024
to see stock.
Add to basket

Description

Description

  • Octagonal shape
  • For vessels with slightly uneven bottoms.
  • Pivot ring reduces the contact surface which enables the stirrer to adopt the optimum position.
Specifications

Specifications

9.5 mm
PTFE
51 mm
10
For vessels with slightly uneven bottoms
Octagon
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers