missing translation for 'onlineSavingsMsg'
Learn More

Octafluoronaphthalene, 96%

Product Code. 10571881 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10571881 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10571881 Supplier Thermo Scientific Acros Supplier No. 291530010
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 313-72-4 | C10F8 | 272.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 313-72-4
CAS Min % 95.0
CAS Max % 100.0
Melting Point 85.0°C to 90.0°C
Color White
Boiling Point 209.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C10F8
MDL Number MFCD00014307
Quantity 1 g
Beilstein 05, 1658
Synonym octafluoronaphthalene, perfluoronaphthalene, naphthalene, octafluoro, octafluoronapthalene, octafiuoronaphthalene, acmc-1cr7y, octafluoronaphthalene 1g, octafluoronaphthalene radical anion
Solubility Information Solubility in water: insoluble.
InChI Key JDCMOHAFGDQQJX-UHFFFAOYSA-N
SMILES C12=C(C(=C(C(=C1F)F)F)F)C(=C(C(=C2F)F)F)F
IUPAC Name 1,2,3,4,5,6,7,8-octafluoronaphthalene
Molecular Weight (g/mol) 272.09
PubChem CID 67564
Formula Weight 272.09
Percent Purity 96%
Physical Form Crystals or Needles
Chemical Name or Material Octafluoronaphthalene, 96%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.