missing translation for 'onlineSavingsMsg'
Learn More

Octafluoro-4,4'-biphenol, TRC

Product Code. 30425330
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30425330 100 mg -
30418544 500 mg 500mg
30425110 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30425330 Supplier Toronto Research Chemicals Supplier No. TRCO273108100MG
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2200-70-6
Molecular Formula C12H2O2F8
Molecular Weight (g/mol) 330.13
IUPAC Name 2,3,5,6-tetrafluoro-4-(2,3,5,6-tetrafluoro-4-hydroxyphenyl)phenol
SMILES Oc1c(F)c(F)c(c(F)c1F)c2c(F)c(F)c(O)c(F)c2F

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C12H2F8O2/c13-3-1(4(14)8(18)11(21)7(3)17)2-5(15)9(19)12(22)10(20)6(2)16/h21-22H
Formula Weight 329.9927
Chemical Name or Material Octafluoro-4,4'-biphenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.