missing translation for 'onlineSavingsMsg'
Learn More

Octafluoro-1,4-diiodobutane, 98%

Product Code. 10664185 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10664185 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10664185 Supplier Thermo Scientific Acros Supplier No. 426320050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 375-50-8 | C4F8I2 | 453.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 375-50-8
CAS Min % 97.5
CAS Max % 100.0
Melting Point -9.0°C
Color Colorless to Yellow or Pink
Density 2.4740g/mL
Boiling Point 150.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C4F8I2
Linear Formula CF2(I)(CF2)2CF2I
Refractive Index 1.4280 to 1.4320 (20°C, 589nm)
MDL Number MFCD00042263
Quantity 5 g
Specific Gravity 2.474
Synonym 1,4-diiodooctafluorobutane, octafluoro-1,4-diiodobutane, 1,4-diiodoperfluorobutane, perfluoro-1,4-diiodobutane, 1,4-diiodo octafluorobutane, butane, 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodo, acmc-1aigi, i cf2 4i, 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodo-butane
InChI Key JILAKKYYZPDQBE-UHFFFAOYSA-N
SMILES FC(F)(I)C(F)(F)C(F)(F)C(F)(F)I
IUPAC Name 1,1,2,2,3,3,4,4-octafluoro-1,4-diiodobutane
Molecular Weight (g/mol) 453.84
PubChem CID 78993
Formula Weight 453.83
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Octafluoro-1, 4-diiodobutane
Show More Show Less
compliance-icons
Product Identifier
  • Octafluoro-1,4-diiodobutane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.