missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecylamine, 90%, tech.
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
Unit Size:
1kg
500g
5kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 88.0 |
| CAS Max % | 100.0 |
| Melting Point | 50.0°C to 60.0°C |
| Color | White |
| Boiling Point | 232.0°C (32.0 mmHg) |
| Flash Point | 148°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 88% min. (GC) |
| Packaging | Plastic drum |
| Linear Formula | CH3(CH2)17NH2 |
| Show More |
Safety and Handling
Product Identifier
- Octadecylamine
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- ACUTE AQUATIC Acute 1
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H315-Causes skin irritation.
- H373-May cause damage to organs.
- H411-Toxic to aquatic life with long lasting effects.
- H304-May be fatal if swallowed and enters airways.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction