missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Octadecanoic-9,9,10,10-d4 Acid, CDN
Specifications
Specifications
| Chemical Name or Material | Octadecanoic-9,9,10,10-D4 Acid |
| CAS | 130048-57-6 |
| Molecular Formula | C18 D4 H32 O2 |
| Quantity | 0.1 g |
| Synonym | Octadecanoic-9,9,10,10-d4 acid (9 CI, ACI), Stearic-9,9,10,10-d4 acid (5 CI), 9,9,10,10-Tetradeuterostearic acid, [9,9,10,10-d4]-Stearic acid, Stearic acid-9,9,10,10-d4, Octadecanoic-9,9,10,10-D4 acid |
| InChI Formula | InChI=1 S/C18H36O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20/h2-17H2,1H3,(H,19,20)/i9D2,10D2 |
| SMILES | [2 H]C([2 H])(CCCCCCCC)C([2 H])([2 H])CCCCCCCC(=O)O |
| IUPAC Name | 9,9,10,10-tetradeuteriooctadecanoic acid |
| Molecular Weight (g/mol) | 288.502 |
| Formula Weight | 288.297 g/mol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?