missing translation for 'onlineSavingsMsg'
Learn More

Octacosane, 99+%

Product Code. 10245320 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10245320 5 g Glass bottle 5g
10306680 25 g Glass bottle 25g
10403255 100 g Glass bottle 100g
13458338 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10245320 Supplier Thermo Scientific Acros Supplier No. 129270050
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 630-02-4 | C28H58 | 394.77 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-02-4
Molecular Formula C28H58
Molecular Weight (g/mol) 394.77
MDL Number MFCD00009355
InChI Key ZYURHZPYMFLWSH-UHFFFAOYSA-N
Synonym n-octacosane, n-octcosane, unii-vff49836p8, ch3-ch2 26-ch3, octacosane, analytical standard, ccris 680, octacosane, n, octacosane, heptacosane, methyl, n-octacosane 10g
PubChem CID 12408
ChEBI CHEBI:32943
IUPAC Name octacosane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 61.0°C to 64.0°C
Color White
Density 0.8067g/mL
Boiling Point 440.0°C
Flash Point 227°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)26CH3
Quantity 5 g
Beilstein 01,I,71
Specific Gravity 0.8067
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, chloroform
Formula Weight 394.76
Percent Purity 99+%
Physical Form Shiny Flakes or Powder
Chemical Name or Material Octacosane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.