Product Code. 10403255
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Unit Size:
100g
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
unitSize
10403255
100 g
Glass bottle
100g
10245320
5 g
Glass bottle
5g
10306680
25 g
Glass bottle
25g
13458338
500 g
Glass bottle
500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code.
10403255
Supplier
Thermo Scientific Acros
Supplier No.
129271000
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
CAS: 630-02-4 | C28H58 | 394.77 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
CAS
630-02-4
Molecular Formula
C28H58
Molecular Weight (g/mol)
394.77
MDL Number
MFCD00009355
InChI Key
ZYURHZPYMFLWSH-UHFFFAOYSA-N
Synonym
n-octacosane, n-octcosane, unii-vff49836p8, ch3-ch2 26-ch3, octacosane, analytical standard, ccris 680, octacosane, n, octacosane, heptacosane, methyl, n-octacosane 10g
PubChem CID
12408
ChEBI
CHEBI:32943
IUPAC Name
octacosane
SMILES
CCCCCCCCCCCCCCCCCCCCCCCCCCCC
Specifications
CAS Min %
99.0
CAS Max %
100.0
Melting Point
61.0°C to 64.0°C
Color
White
Density
0.8067g/mL
Boiling Point
440.0°C
Flash Point
227°C
Infrared Spectrum
Authentic
Assay Percent Range
99% min. (GC)
Packaging
Glass bottle
Linear Formula
CH3 (CH2 )2 6 CH3
Quantity
100 g
Beilstein
01,I,71
Specific Gravity
0.8067
Solubility Information
Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, chloroform
Formula Weight
394.76
Percent Purity
99+%
Physical Form
Shiny Flakes or Powder
Chemical Name or Material
Octacosane
Show More
Show Less
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?