missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
44475 Polyclonal specifically detects 44475 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | OCT6. |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry-Paraffin |
| Formulation | PBS buffer, 2% sucrose |
| Gene Alias | Oct-6, OCT6POU domain transcription factor SCIP, Octamer-binding protein 6, Octamer-binding transcription factor 6, OTF6, OTF-6, POU class 3 homeobox 1, POU domain class 3, transcription factor 1, POU domain, class 3, transcription factor 1, SCIP |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Rat OCT6 (NP_620193). Peptide sequence CPKPSAHEITGLADSLQLEKEVVRVWFCNRRQKEKRMTPAAGAGHPPMDD |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?