Learn More
Invitrogen™ OCT6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110202
Description
Immunogen sequence: MATTAQYLPR GPGGGAGGTG PLMHPDAAAA AAAAAAAERL HAGAAYREVQ KLMHHEWLGA GAGHPVGLAH PQWLPTGGGG GGDWAGGPHL EHGKAGGGGT GRADDGGGGG GFHARLVHQG AAHAGAAWAQ GSTAHHLGPA MSPSPGASGG HQPQPLGLYA QAAYPGGGGG GLAGMLAAGG GGAGPGLHHA LHEDGHEAQL EPSPPPHLGA HGHAHGHAHA GGLHAAAAHL HPGAGGGGSS VGEHSDEDAP SSDDLEQFAK.
POU3F1 is a member of the POU domain family of proteins, and regulates events during neurogenesis and myelination.
Specifications
| OCT6 | |
| Polyclonal | |
| Unconjugated | |
| POU3F1 | |
| 6-Oct; LOW QUALITY PROTEIN: POU domain, class 3, transcription factor 1; Oct6; Oct-6; octamer binding factor oct6; octamer-binding protein 6; Octamer-binding transcription factor 6; octoamer transcription factor-6; octomer-6; Otf6; OTF-6; POU class 3 homeobox 1; POU domain transcription factor SCIP; POU domain, class 3, transcription factor 1; POU3F1; Scip; suppressed cAMP-inducible POU; Test1; Testes-1; Tst1; Tst-1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 18991, 192110 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.98 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P20267, P21952 | |
| POU3F1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human POU3F1 (NP_0026903). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.