missing translation for 'onlineSavingsMsg'
Learn More

OCT4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen OCT4
Concentration 0.36 mg/mL
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16808223
View Documents
Proteintech
11263-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16898213
View Documents
Proteintech
11263-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Oct4 is a maternally expressed octamer-binding protein that was the first transcription factor described for the early stages of development. Oct4 is one of the four Yamaka factors, necessary for embryonic stem cell pluripotency. Pluripotent stem (iPS) cells, can be created by expressing Oct4 and the transcription factors Sox2, Klf4 and Lin28 along with c-Myc in mouse fibroblasts. More recently, experiments have demonstrated that iPS cells could be generated using expression plasmids expressing Oct4, Sox2, KlfF4 and c-Myc, eliminating the need for virus introduction, thereby addressing a safety concern for potential use of iPS cells in regenerative medicine. A translocation of this gene with the Ewing′s sarcoma gene, t(6;22)(p21;q12), has been linked to tumor formation.
Specifications

Specifications

OCT4
Western Blot
Unconjugated
Rabbit
Rat, Mouse, Human
P20263, Q01860
18999, 294562, 5460
OCT4 Fusion Protein Ag1794
Primary
-20°C
POU5F1
0.36 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Oct 3, Oct 4, OCT3, OCT3/4, OCT4, Octamer binding protein 3, Octamer binding protein 4, OTF 3, OTF3, OTF4, POU class 5 homeobox 1, POU5F1
Pou5f1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.