missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT4 Rabbit anti-Human, Clone: OCT4/6875R, Novus Biologicals™
Rabbit Monoclonal Antibody
£219.00 - £411.00
Specifications
| Antigen | OCT4. |
|---|---|
| Clone | OCT4/6875R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18077143
|
Novus Biologicals
NBP3-13745-20UG |
20 μg |
£219.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18366695
|
Novus Biologicals
NBP3-13745-100UG |
100 μg |
£411.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OCT4 Monoclonal antibody specifically detects OCT4. in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| OCT4. | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators | |
| 10 mM PBS with 0.05% BSA | |
| class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein | |
| Recombinant fragment of human OCT4 protein (around aa 1-134) (exact sequence is proprietary) (Uniprot: Q01860) | |
| Primary | |
| Store at 4°C. |
| OCT4/6875R | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q01860 | |
| 5460 | |
| IgG κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title