missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£181.00 - £265.00
Specifications
| Antigen | OCT4 |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:300-1:10000 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18691816
|
Novus Biologicals
NBP3-05882-100ul |
100 μg |
£265.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18663616
|
Novus Biologicals
NBP3-05882-50ul |
50 μg |
£181.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OCT4 Polyclonal antibody specifically detects OCT4 in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| OCT4 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators | |
| 0.2 um filtered solution in PBS | |
| 5460 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunocytochemistry/ Immunofluorescence 1:300-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human OCT4. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title