missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT4 Antibody (A3-C11-R), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-31983
This item is not returnable.
View return policy
Description
OCT4 Monoclonal antibody specifically detects OCT4 in Human samples. It is validated for Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| OCT4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Immunocytochemistry/Immunofluorescence 1:100 | |
| class 5, transcription factor 1, MGC22487, Oct-3, OCT3Oct4, Oct-4, Octamer-binding protein 3, otc-4, OTF3POU domain class 5, transcription factor 1, OTF4, POU class 5 homeobox 1, POU-type homeodomain-containing DNA-binding protein | |
| Recombinant protein within full length human OCT4 protein. (Uniprot: Q01860) | |
| 100 μL | |
| Cellular Markers, Chromatin Research, Glia Markers, Neuroscience, Oligodendrocyte Cell Markers, Pancreatic Stem Cell Markers, Stem Cell Markers, Stem Cells, Transcription Factors and Regulators | |
| 5460 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| A3-C11-R | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction