missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OCT2 Rabbit anti-Human, Clone: S02-5E7, Novus Biologicals™
Rabbit Monoclonal Antibody
£222.00 - £451.00
Specifications
| Antigen | OCT2. |
|---|---|
| Clone | S02-5E7 |
| Dilution | Western Blot |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18313297
|
Novus Biologicals
NBP3-14869-25UL |
25 μg |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18336255
|
Novus Biologicals
NBP3-14869-100UL |
100 μg |
£451.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
OCT2 Monoclonal antibody specifically detects OCT2 in Human samples. It is validated for Western BlotSpecifications
| OCT2. | |
| Western Blot | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| homeobox protein, Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2, OCT2Oct-2, Octamer-binding protein 2, Octamer-binding transcription factor 2, OTF-2, OTF2POU domain class 2, transcription factor 2, POU class 2 homeobox 2, POU domain, class 2, transcription factor 2 | |
| A synthetic peptide of human OCT2 (Uniprot#: P09086) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S02-5E7 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| 5452 | |
| IgG | |
| Affinity purified | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title