missing translation for 'onlineSavingsMsg'
Learn More

OCT2 Antibody (OCT2/7073R), Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18339886 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18339886 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18339886 Supplier Novus Biologicals Supplier No. NBP314146AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

OCT2 Monoclonal antibody specifically detects OCT2 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCT2.
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone OCT2/7073R
Conjugate Alexa Fluor 405
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias homeobox protein, Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2, OCT2Oct-2, Octamer-binding protein 2, Octamer-binding transcription factor 2, OTF-2, OTF2POU domain class 2, transcription factor 2, POU class 2 homeobox 2, POU domain, class 2, transcription factor 2
Host Species Rabbit
Immunogen Recombinant fragment of human OCT2 protein (around aa 112-297) (exact sequence is proprietary) (Uniprot: P09086)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 5452
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.