missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
OCT2 Monoclonal antibody specifically detects OCT2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
Specifications
| Antigen | OCT2 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Classification | Monoclonal |
| Clone | Oct2/2136 |
| Concentration | 0.2 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array |
| Formulation | 10 mM PBS with 0.05% BSA |
| Gene Accession No. | P09086 |
| Gene Alias | homeobox protein, Lymphoid-restricted immunoglobulin octamer-binding protein NF-A2, OCT2Oct-2, Octamer-binding protein 2, Octamer-binding transcription factor 2, OTF-2, OTF2POU domain class 2, transcription factor 2, POU class 2 homeobox 2, POU domain, class 2, transcription factor 2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?