missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OCT1 (POU2F1) Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA519302
This item is not returnable.
View return policy
Description
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.
OCT1 (POU2F1) is a ubiquitously expressed transcription factor that binds to the octamer motif (5'-ATTTGCAT-3') and activates the promoters of the genes for some small nuclear RNAs (snRNA) and of genes such as those for histone H2B and immunoglobulins. It modulates transcription transactivation by NR3C1, AR and PGR. In case of human herpes simplex virus (HSV) infection, POU2F1 forms a multiprotein-DNA complex with the viral transactivator protein VP16 and HCFC1 thereby enabling the transcription of the viral immediate early genes.
Specifications
| OCT1 (POU2F1) | |
| Polyclonal | |
| Unconjugated | |
| POU2F1 | |
| 2810482H01Rik; NF-A1; OCT1; oct-1; oct-1B; octamer binding protein 1; octamer-binding protein 1; Octamer-binding transcription factor 1; Octamer-binding transcription factor-1; Otf1; OTF-1; POU class 2 homeobox 1; POU domain, class 2, transcription factor 1; Pou2f1; ubiquitous transcription factor | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 5451 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P14859 | |
| POU2F1 | |
| Synthetic peptide sequence (NVQSKSNEESGDSQQ) corresponding to the internal amino acids of POU2F1 (aa 69-84). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction