Learn More
CELLPATH OCT EMBEDDING MATRIX
Product Code. 30226281
Change view
Click to view available options
Form:
Liquid
Unit Size:
125mL
Product Code. Form Unit Size
30226281 Liquid 125mL
1 options
Product Code. 30226281 Supplier CELLPATH Supplier No. KMA010000A
  • ONLINE EXCLUSIVE PRICE
    £21.21
    £10.00 / 125mL
    Save £11.21
    52% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
OCT cryo microtomy embedding medium is designed for use at -20°C to -30°C. The medium freezes rapidly with minimal damage from crystal formation and is an ideal support medium for cutting tissue sections.
  • Key Benefits:
  • Liquid level visible in easy to use dispenser
  • Miscible with water to facilitate easy removal of tissue from the block holder
  • Easily dissolved from the microscope slide before staining
  • High viscosity to ensure strong adhesion of the tissue to the holder allowing correct orientation of the specimen while the medium is liquid
  • Freezes rapidly with minimal damage by crystal formation
  • Can be used as an infiltrating media for tissues that tend to shatter on cutting, such as brain
Specifications
Product Type Embedding Media
Form Liquid