missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oct-4B Antibody (OTI7H1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73141PE
This item is not returnable.
View return policy
Description
Oct-4B Monoclonal antibody specifically detects Oct-4B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Oct-4B | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 5460 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| OTI7H1 | |
| Western Blot, Immunocytochemistry/ Immunofluorescence | |
| OCT3,Oct-3,OCT4,Oct-4,OCT4,Oct3/4,OTF3,OTF-3,OTF4,POU domain protein,POU domain, class 5, transcription factor 1,POU5F1 | |
| Full length human recombinant protein of human POU5F1(NP_002692) produced in HEK293T cell. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction