missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Oct-2 Recombinant Rabbit Monoclonal Antibody (K01_2N70)

Product Code. 30829608
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829608 50 μL
30829277 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829608 Supplier Invitrogen™ Supplier No. MA565612

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: NPSPQGSHSA IGLSGLN.

OCT-2 is a transcription factor belonging to the POU homeo-domain family that binds to the Ig gene octamer sites regulating B cell specific genes. OCT-2 protein expression is not restricted to B cells, although expression levels are much higher in these cells. OCT-2 expression is reported to be significantly greater in germinal center derived lymphomas, although other B cell lymphomas also display high levels of expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oct-2
Applications Western Blot
Classification Recombinant Monoclonal
Clone K01_2N70
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene POU2F2
Gene Accession No. P09086
Gene Alias 2-Oct; homeobox protein; lymphoid-restricted immunoglobulin octamer-binding protein NF-A2; OCT2; Oct-2; Oct2a; Oct2b; octamer-binding protein 2; Octamer-binding transcription factor 2; OTF2; OTF-2; POU class 2 homeobox 2; POU domain, class 2, transcription factor 2; Pou2f2
Gene Symbols POU2F2
Host Species Rabbit
Immunogen A synthetic peptide of human Oct-2 (aa 447-463).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5452
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.