missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Oct-2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17896232
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17896232 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17896232 Supplier Invitrogen™ Supplier No. PA5142573

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.

OCT-2 is a transcription factor belonging to the POU homeo-domain family that binds to the Ig gene octamer sites regulating B cell specific genes. OCT-2 protein expression is not restricted to B cells, although expression levels are much higher in these cells. OCT-2 expression is reported to be significantly greater in germinal center derived lymphomas, although other B cell lymphomas also display high levels of expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oct-2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene POU2F2
Gene Accession No. P09086, Q00196
Gene Alias 2-Oct; homeobox protein; lymphoid-restricted immunoglobulin octamer-binding protein NF-A2; OCT2; Oct-2; Oct2a; Oct2b; octamer-binding protein 2; Octamer-binding transcription factor 2; OTF2; OTF-2; POU class 2 homeobox 2; POU domain, class 2, transcription factor 2; Pou2f2
Gene Symbols POU2F2
Host Species Goat
Immunogen Peptide with sequence KQGLDSPSEHTDTER-C (aa 21-35).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18987, 5452
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.