missing translation for 'onlineSavingsMsg'
Learn More

Oct-1-ene, Pure, Fisher Chemical

Product Code. 10418990 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10418990 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10418990 Supplier Fisher Chemical Supplier No. O/0105/PB05

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Packaging Plastic Coated Glass bottle
CAS 111-66-0
Vapor Pressure 13mmHg at 20°C
Viscosity 0.38 St at 40°C
Vapor Density 3.9
Physical Form Liquid
Molecular Formula C8H16
MDL Number 9548
Melting Point -102°C
Boiling Point 121°C
InChI Key KWKAKUADMBZCLK-UHFFFAOYSA-N
SMILES CCCCCCC=C
IUPAC Name oct-1-ene
Molecular Weight (g/mol) 112.216
PubChem CID 8125
Quantity 100 mL
ChEBI CHEBI:46708
Formula Weight 112.21g/mol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Octene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H410-Very toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.