missing translation for 'onlineSavingsMsg'
Learn More

OCRL Antibody [Biotin], R&D Systems™

Product Code. 30500573 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30500573 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30500573 Supplier Novus Biologicals Supplier No. NBP337944B
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

OCRL Polyclonal antibody specifically detects OCRL in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCRL
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Biotin
Formulation PBS
Gene Alias EC 3.1.3.36, INPP5F, LOCR, Lowe oculocerebrorenal syndrome protein, NPHL2, OCRL1inositol polyphosphate 5-phosphatase OCRL-1, oculocerebrorenal syndrome of Lowe, phosphatidylinositol polyphosphate 5-phosphatase
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human OCRL (NP_001578.2).,, Sequence:, MEPPLPVGAQPLATVEGMEMKGPLREPCALTLAQRNGQYELIIQLHEKEQHVQDIIPINSHFRCVQEAEETLLIDIASNSGCKIRVQGDWIRERRFEIPDEEHCLKFLSAVLAAQKAQSQLLVPEQKDSSSWYQKLDTKDKPSVFSGLLGFEDNFSSMNLDKKINSQNQPTGIHREPPPPPFSVNKMLPREKEASNKEQPKVTNTMRKLFVPNTQSGQRE
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Golgi Apparatus Markers
Primary or Secondary Primary
Gene ID (Entrez) 4952
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.