missing translation for 'onlineSavingsMsg'
Learn More

Oclacitinib Maleate, TRC

Product Code. 30135111
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135111 5 mg 5mg
30133205 1 mg 1mg
30009594 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135111 Supplier Toronto Research Chemicals Supplier No. TRCO1700205MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1640292-55-2
Molecular Formula C19H27N5O6S
Molecular Weight (g/mol) 453.51
SMILES CNS(=O)(=O)C[C@@H]1CC[C@H](CC1)N(C)c2ncnc3[nH]ccc23.OC(=O)\C=C/C(=O)O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C15H23N5O2S.C4H4O4/c1-16-23(21,22)9-11-3-5-12(6-4-11)20(2)15-13-7-8-17-14(13)18-10-19-15;5-3(6)1-2-4(7)8/h7-8,10-12,16H,3-6,9H2,1-2H3,(H,17,18,19);1-2H,(H,5,6)(H,7,8)/b;2-1-
Chemical Name or Material Oclacitinib Maleate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.