missing translation for 'onlineSavingsMsg'
Learn More

OCIAD2 Antibody, FITC, Novus Biologicals™

Product Code. 18429790 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18429790 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18429790 Supplier Novus Biologicals Supplier No. NBP177085F

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OCIAD2 Polyclonal antibody specifically detects OCIAD2 in Human, Mouse, Rat samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCIAD2
Applications ELISA
Classification Polyclonal
Conjugate FITC
Formulation PBS
Gene Alias MGC45416, OCIA domain containing 2, OCIA domain-containing protein 2, ovarian carcinoma immunoreactive antigen, ovarian carcinoma immunoreactive antigen-like protein
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the amino terminus of human OCIAD2. The immunogen is located within the first 50 amino acids of OCIAD2. Amino Acid Squence: SARGNQDKDAHFPPPSK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 132299
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.