missing translation for 'onlineSavingsMsg'
Learn More

OCIAD1 Antibody, Novus Biologicals™

Product Code. 18361779 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18361779 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18361779 Supplier Novus Biologicals Supplier No. H00054940B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

OCIAD1 Polyclonal antibody specifically detects OCIAD1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCIAD1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500
Formulation PBS (pH 7.4)
Gene Accession No. AAH03409
Gene Alias Asrij, MGC111072, OCIA domain containing 1, OCIA domain-containing protein 1, OCIAFLJ20455, Ovarian carcinoma immunoreactive antigen, TPA018
Host Species Mouse
Immunogen OCIAD1 (AAH03409, 1 a.a. - 245 a.a.) full-length human protein. MNGRADFREPNAEVPRPIPHIGPDYIPTEEERRVFAECNDESFWFRSVPLAATSMLITQGLISKGILSSHPKYGSIPKLILACIMGYFAGKLSYVKTCQEKFKKLENSPLGEALRSGQARRSSPPGHYYQKSKYDSSVSGQSSFVTSPAADNIEMLPHYEPIPFSSSMNESAPTGITDHIVQGPDPNLEESPKRKNITYEELRNKNRESYEVSLTQKTDPSVRPMHERVPKKEVKVNKYGDTWDE
Purification Method IgG purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 54940
Target Species Human, Mouse
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.