missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OCEL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559851
This item is not returnable.
View return policy
Description
Immunogen sequence: HKAKTKKIVF EDELLSQALL GAKKPIGAIP KGHKPRPHPV PDYELKYPPV SSERERSRYV AVFQDQYGEF LELQH Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 60%.
Specifications
| OCEL1 | |
| Polyclonal | |
| Unconjugated | |
| OCEL1 | |
| 9430098E02Rik; AI326022; AI593544; FWP009; occludin/ELL domain containing 1; occludin/ELL domain-containing protein 1; OCEL1; RGD1563236; RGD1565407; S863-9 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79629 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H607 | |
| OCEL1 | |
| Recombinant protein corresponding to Human OCEL1. Recombinant protein control fragment (Product #RP-98903). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction