missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
OCAM/NCAM2 Polyclonal antibody specifically detects OCAM/NCAM2 in Human samples. It is validated for ELISA
Specifications
Specifications
| Antigen | OCAM/NCAM2 |
| Classification | Polyclonal |
| Host Species | Rabbit |
| Regulatory Status | RUO |
| Gene ID (Entrez) | 4685 |
| Target Species | Human |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?