missing translation for 'onlineSavingsMsg'
Learn More

OCA2 Antibody (1A11), Novus Biologicals™

Product Code. 18326408 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18326408 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18326408 Supplier Novus Biologicals Supplier No. H00004948M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

OCA2 Monoclonal antibody specifically detects OCA2 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen OCA2
Applications ELISA
Classification Monoclonal
Clone 1A11
Conjugate Unconjugated
Dilution ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH12097
Gene Alias BEY, BEY1, BEY2, D15S12BOCA, EYCL, EYCL2, eye color 2 (central brown), eye color 3 (brown), hair color 3 (brown), HCL3, Melanocyte-specific transporter protein, oculocutaneous albinism II, oculocutaneous albinism II (pink-eye dilution (murine) homolog), oculocutaneous albinism II (pink-eye dilution homolog, mouse), P protein, PED, PEYCL3, Pink-eyed dilution protein homolog, SHEP1, total brown iris pigmentation
Host Species Mouse
Immunogen OCA2 (AAH12097, 201 a.a. ~ 300 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. GKLWQLLALSPLENYSVNLSSHVDSTLLQVDLAGALVASGPSRPGREEHIVVELTQADALGSRWRRPQQVTHNWTVYLNPRRSEHSVMSRTFEVLTRETV
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4948
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.