missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OBSCN Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97423V
This item is not returnable.
View return policy
Description
OBSCN Polyclonal specifically detects OBSCN in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| OBSCN | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ARHGEF30, DKFZp666E245, EC 2.7.11.1, FLJ14124, KIAA1556MGC120412, KIAA1639MGC138590, MGC120409, MGC120410, MGC120411, obscurin, obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF, obscurin, myosin light chain kinase, Obscurin-MLCK, Obscurin-myosin light chain kinase, obscurin-RhoGEF, UNC89 | |
| Produced in rabbits immunized with E. coli-derived Human OBSCN fragment. | |
| 0.1 mL | |
| Protein Kinase | |
| 84033 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction