missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OBP2B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£359.00
Specifications
| Antigen | OBP2B |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
OBP2B Polyclonal specifically detects OBP2B in Human samples. It is validated for Western Blot.Specifications
| OBP2B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| hOBPIIb, MGC119022, OBPIIb, odorant binding protein 2B, odorant-binding protein 2b, odorant-binding protein IIb, UNQ653/PRO1283 | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human OBP2B. Peptide sequence RKLMYLQELPRRDHYIFYCKDQHHGGLLHMGKLVGRNSDTNREALEEFKK | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 29989 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title