missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
OBP2B Polyclonal antibody specifically detects OBP2B in Human samples. It is validated for ELISA, Immunofluorescence
Specifications
Specifications
| Antigen | OBP2B |
| Applications | ELISA, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | DyLight 650 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | hOBPIIb, MGC119022, OBPIIb, odorant binding protein 2B, odorant-binding protein 2b, odorant-binding protein IIb, UNQ653/PRO1283 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human OBP2B (NP_055396.1; Met1-His170). |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?