missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OBP2B Antibody (002), Alexa Fluor™ 647, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90310AF647
This item is not returnable.
View return policy
Description
OBP2B Monoclonal specifically detects OBP2B in Human samples. It is validated for ELISA.
Specifications
| OBP2B | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 29989 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 2 | |
| ELISA | |
| hOBPIIb, MGC119022, OBPIIb, odorant binding protein 2B, odorant-binding protein 2b, odorant-binding protein IIb, UNQ653/PRO1283 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human OBP2B (NP_055396.1; Met1-His170). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction