missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OBP2A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563220
This item is not returnable.
View return policy
Description
Immunogen sequence: RNWCSTRDSR RRTFSRPCRR EAAFSNTRQP PGLHLQSPPY HQTQSPDHLD LPSSHDPSLL PP Highest antigen sequence identity to the following orthologs: Mouse - 29%, Rat - 31%.
This gene encodes a small extracellular protein belonging to the lipocalin superfamily. The protein is thought to transport small, hydrophobic, volatile molecules or odorants through the nasal mucus to olfactory receptors, and may also function as a scavenger of highly concentrated or toxic odors. The protein is expressed as a monomer in the nasal mucus, and can bind diverse types of odorants with a higher affinity for aldehydes and fatty acids. This gene and a highly similar family member are located in a cluster of lipocalin genes on chromosome 9. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.
Specifications
| OBP2A | |
| Polyclonal | |
| Unconjugated | |
| OBP2A | |
| hOBPIIa; LCN13; OBP; OBP2A; OBP2C; OBPIIa; odorant binding protein 2A; odorant-binding protein 2a; Odorant-binding protein IIa; putative odorant-binding protein 2c | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 29991 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NY56 | |
| OBP2A | |
| Recombinant protein corresponding to Human OBP2A. Recombinant protein control fragment (Product #RP-101826). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction