missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OBP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144567
This item is not returnable.
View return policy
Description
OBP Polyclonal Antibody for Western Blot, ELISA
This protein is found in nasal epithelium and it binds a wide variety of chemical odorants.
Specifications
| OBP | |
| Polyclonal | |
| Unconjugated | |
| OBP | |
| 1110006M06Rik; AI462538; Obph1; ORP5; ORP-5; Osbp2; Osbpl5; OSBP-related protein 5; oxysterol binding protein 2; oxysterol binding protein homolog 1; oxysterol binding protein-like 5; oxysterol-binding protein homolog 1; oxysterol-binding protein-like protein 5; oxysterol-binding protein-related protein 5 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 532690, 785083 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 | |
| P07435 | |
| OBP, Osbpl5 | |
| Recombinant Bos taurus Odorant-binding protein (1-159aa). | |
| 100 μL | |
| Primary | |
| Bovine | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction