Learn More
Motic™ Objective, Plan Phase PL Ph 40X/0.65/S
Product Code. 15485122
Change view
Click to view available options
Magnification Power:
40X
Unit Size:
Each
Product Code. Magnification Power unitSize
15485122 40X Each
1 options
Product Code. 15485122 Supplier Motic™ Supplier No. 12220260
Estimated Shipment: 22-09-2026
Add to basket
Add to basket
Designed with field curvature correction and an integrated phase plate for contrast modulation. Motic™ Objective, Plan Phase PL Ph 40X/0.65/S provides a flattened field of view and high-contrast images of transparent specimens.
  • Plan style for field curvature correction.
  • Integrated phase plate for contrast modulation.
Specifications
Type Objective
Description NA: 0.65/S
For Use With (Equipment) B1-223ASC, B3-223PL, DMWB1-223ASC Microscopes
Magnification Power 40X