Learn More
Motic™ Objective Lens for SMZ160
Product Code. 17817285
Click to view available options
Magnification Power:
0.35X
0.5X
0.75X
1.5X
2X
Unit Size:
Each
Product Code. 17817285
Brand: Motic™ 1101001708271
Only null left
Add to basket
Add to basket
The Objective Lens is compatible with the SMZ-160 microscope.
  • Objective Lens compatible with SMZ160.
  • Not compatible with standard stands.
  • Available in sizes 0.35x with WD = 287 mm, 0.5x with WD = 165 mm, 0.75x with WD = 120 mm, 1.5x with WD = 47 mm and 2.0 with WD = 30 mm.
Specifications
Type SMZ160 Objective
Focus Distance 120 mm
For Use With (Application) Reduce zoom range of SMZ-160
For Use With (Equipment) SMZ-160
Magnification Power 0.75X
Warranty 5 years
Working Distance 120 mm