Learn More
Leica Microsystems Objective, HI-Plan
Product Code. p-8008330
Click to view available options
Magnification Power:
100X
10X
Unit Size:
Each
Product Code. 11964419
Brand: Leica Microsystems 11506230
This item requires a price update prior to being ordered.
Request a Price for this item here.
Economical, high quality microscope objective designed for visual fields up to 20mm. Leica Microsystems™ Objective HI-Plan features good chromatic correction at two wavelengths, flatness over the whole field of view and sharp image edges.
The Leica DM1000 is the ideal instrument for biological, medical and clinical laboratories. The instrument can be configured to meet specific requirements by choosing from a selection of observation tubes, ergonomic modules, objectives and adjustable controls. Please note a complete system comprises the main microscope unit, an observation tube and either the brightfield or phase contrast outfit. Fluorescence outfits also require the brightfield or phase contrast outfit to be ordered.
Specifications
Type Objective HI-Plan
Magnification Power 10X
Quantity 1 Ea.