Learn More
Motic™ Objective, Achromatic EA 40X/0.65/S
Product Code. 15465102
Click to view available options
Magnification Power:
40X
Unit Size:
Each
Product Code. 15465102
Brand: Motic™ 12180164
Only null left
Add to basket
Add to basket
Designed with chromatic aberration correction for 2 colors. Motic™ Objective, Achromatic EA 40X/0.65/S is spherically corrected for one color.
  • Achromatic.
  • Chromatic aberration correction for 2 colors.
  • Spherical correction for 1 color.
Specifications
Type Achromat Objective
Description NA: 0.65S
Focus Distance 0.30 mm WD
For Use With (Equipment) PM Series, DM1802-A, SERIES 18, Series 28 Microscopes
Magnification Power 40X