Learn More
Motic™ Objective, Achromatic 60X/0.85/S
Product Code. 15425092
Click to view available options
Magnification Power:
60X
Unit Size:
Each
Product Code. 15425092
Brand: Motic™ 1101001701171
Only null left
Add to basket
Add to basket
Designed with chromatic aberration correction for 2 colors. Motic™ Objective, Achromatic 60X/0.85/S is spherically corrected for one color.
  • Achromatic.
  • Chromatic aberration correction for 2 colors.
  • Spherical correction for 1 color.
Specifications
Type Achromat Objective
Description NA: 0.85/S
For Use With (Equipment) SFC-100, DM-111, F-1110 Microscopes
Magnification Power 60X