Learn More
Leica Microsystems Objective, 0.63X, C-Mount, M-Series Stereomicroscopes
Click to view available options
Magnification Power:
0.63X
Unit Size:
Each
Description
- Designed to accept video coupling (C-mount).
- For M-series stereomicroscopes.
- Works best with 2/3inch or smaller camera chips.
Specifications
Specifications
| Type | Objective |
| Magnification Power | 0.63X |