Learn More
Zeiss™ Objective
Product Code. 16407560
Change view
Click to view available options
Type:
Objective LD A-Plan 40x/0.55 Ph2 M27
Objective N-Achroplan 10x/0.25 M27
Magnification Power:
10X
40X
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Magnification Power Unit Size
16407560 Objective N-Achroplan 10x/0.25 M27 10X Each
13488938 Objective LD A-Plan 40x/0.55 Ph2 M27 40X Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16407560 Supplier Zeiss™ Supplier No. 4209409901000
  • ONLINE EXCLUSIVE PRICE
    £500.85
    £459.00 / Pack of 1
    Save £41.85
    8% Off
Estimated Shipment: 29-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Accessories for Zeiss Microscopes
For use with a variety of applications
Specifications
Type Objective N-Achroplan 10x/0.25 M27
Description Numerical Aperture: 0.25
Focus Distance 6.5 mm
For Use With (Equipment) Axio Imager, Axio Lab, Axio Observer, Axio Scan, Axio Vert and Axio Scope Microscopes
Magnification Power 10X
Quantity 1 Ea.