Learn More
Zeiss™ Objective
Click to view available options
Type:
Objective LD A-Plan 40x/0.55 Ph2 M27
Objective N-Achroplan 10x/0.25 M27
Magnification Power:
10X
40X
Unit Size:
Each
Description
For use with a variety of applications
Specifications
Specifications
| Type | Objective LD A-Plan 40x/0.55 Ph2 M27 |
| Description | Numerical Aperture: 0.55 |
| Focus Distance | 2 mm |
| For Use With (Equipment) | Axio Observer and Axio Vert Microscopes |
| Magnification Power | 40X |
| Quantity | 1 Ea. |