missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OBFC2A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16719-1-AP-20UL
This item is not returnable.
View return policy
Description
Single-stranded DNA.Specifications
| OBFC2A | |
| Polyclonal | |
| Unconjugated | |
| NABP1 | |
| hSSB2, OBFC2A, Sensor of ssDNA subunit B2, SOSS B2, SOSS complex subunit B2, SSB2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 109019, 363227, 64859 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5FVP2, Q8BGW5, Q96AH0 | |
| Nabp1 | |
| OBFC2A Fusion Protein Ag10158 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction