missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OBFC2A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-26640
This item is not returnable.
View return policy
Description
OBFC2A Polyclonal specifically detects OBFC2A in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications
| OBFC2A | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| DKFZp667M1322, FLJ13624, FLJ22833, hSSB2, MGC111163, oligonucleotide/oligosaccharide-binding fold containing 2A, Oligonucleotide/oligosaccharide-binding fold-containing protein 2A, Sensor of single-strand DNA complex subunit B2, Sensor of ssDNA subunit B2, SOSS complex subunit B2, SOSS-B2, SSB2Single-stranded DNA-binding protein 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Immunoprecipitation, Western Blot | |
| 1.0 mg/mL | |
| Immunoprecipitation 5-10 ug/mg lysate, Western Blot (Negative) | |
| NP_001026886.1 | |
| NABP1 | |
| The immunogen recognized by this antibody maps to a region between residue 150 to 200 of human oligonucleotide/oligosaccharide-binding fold containing 2A using the numbering given in entry NP_001026886.1 (GeneID 64859). | |
| 100 μL | |
| DNA replication Transcription Translation and Splicing | |
| 64859 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction