missing translation for 'onlineSavingsMsg'
Learn More

OBFC2A Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18402336 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402336 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18402336 Supplier Novus Biologicals Supplier No. NBP126640JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OBFC2A Polyclonal antibody specifically detects OBFC2A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen OBFC2A
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias DKFZp667M1322, FLJ13624, FLJ22833, hSSB2, MGC111163, oligonucleotide/oligosaccharide-binding fold containing 2A, Oligonucleotide/oligosaccharide-binding fold-containing protein 2A, Sensor of single-strand DNA complex subunit B2, Sensor of ssDNA subunit B2, SOSS complex subunit B2, SOSS-B2, SSB2Single-stranded DNA-binding protein 2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 150 to 200 of human oligonucleotide/oligosaccharide-binding fold containing 2A using the numbering given in entry NP_001026886.1 (GeneID 64859).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 64859
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.